Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vesicular Glutamate Transporter 3 (VGLUT3), Rabbit anti-Mouse, Human, Rat, Polyclonal, Millipore Sigma™

Catalog No. AB5421II25 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5421II25 25 μg
AB5421II100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB5421II25 Supplier MilliporeSigma Supplier No. AB5421II25UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Vesicular Glutamate Transporter 3 (VGLUT3), Cat. No. AB5421-II, is a highly specific rabbit polyclonal antibody that targets Vesicular glutamate transporter 3 and has been tested foe use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Vesicular glutamate transporter 3 (UniProt Q7TSF2; also known as Solute carrier family 17 member 8, VGluT3) is encoded by the Slc17a8 (also known as Vglut3) gene (Gene ID 266767) in rat species. VGluTs are proton-dependent carriers that are responsible for the upload of the major excitatory neurotransmitter glutamate into synaptic vesicles within presynaptic terminals to allow later regulated release at the synaptic cleft. VGluT3 is a multi-pass synaptic vesicle membrane protein that mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neurons. Rat VGluT3 has 12 helical, 7 cytoplasmic, and 6 vesicular domains. It is structurally similar to VGluT1 and -2, but differs in its distribution pattern. Unlike VGluT1 and 2 that rely on interactions with the endocytic machinery for recycling, but remain almost exclusively axonal, VGluT3 can target to the cell body and dendrites, as well as the axon of certain neurons and is implicated in the retrograde synaptic release of glutamate. VGluT3 displays a very restricted localization and is not present in known glutamatergic neurons, but is found in cholinergic neurons in the caudate-putamen and in serotonergic neurons in the raphe nucleus. VGluT3 is localized over vesicle clusters in both symmetric (inhibitory or modulatory) and asymmetric (excitatory) synapses. (Ref.: Herzog, E et al. (2004). Neuroscience. 123(4):983-1002; Seal, RP et al. (2008). Neuron 57(2); 263-275).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vesicular Glutamate Transporter 3 (VGLUT3)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:500 dilution from a representative lot detected Vesicular Glutamate Transporter 3 (VGLUT3) in PC12 cell lysate.Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected Vesicular Glutamate Transporter 3 (VGLUT3) in rat brain and human brain tissue sections.Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Vesicular Glutamate Transporter 3 (VGLUT3) in PC12 cells.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Solute carrier family 17 member 8;VGluT3
Gene Symbols Slc17a8;Vglut3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of rat Vesicular glutamate transporter 3 (VGluT3).
Molecular Weight of Antigen ∽ 60 kDa observed; 64.76 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects Vesicular glutamate transporter 3(VGluT3). It targets an epitope with in 13 amino acids from the C-terminal region.
Target Species Mouse, Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.