Learn More
TARGETMOL CHEMICALS INC SKLB 1028 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T34656
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SKLB 1028 (Ruserontinib) is a new type of oral multikinase inhibitor of EGFR FLT3 and Abl. SKLB 1028 shows excellent activity in FLT3-driven AML models and considerable potency in CML models containing Abl mutants. purity: 99%
Catalog No. 50-257-6938
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.