Learn More
Vespel™ Needle Kit for TD Tubes, Sold by MilliporeSigma™ Supelco™
Catalog No. 111010169 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
Attaching a Needle to The Inlet of The Tube
Catalog No. For Use With (Application)
11-101-0169 Attaching a Needle to The Inlet of The Tube
1 options
Catalog No. 11-101-0169 Supplier MilliporeSigma Supelco Supplier No. 29023U
Add to Cart
Add to Cart
An accessory designed to assist users of thermal desorption tubes in attaching a needle to the inlet of the tube
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Attaching a Needle to The Inlet of The Tube
Includes Stainless Steel 22 s G Needle (Bevel Point), Stainless Steel Fitting, Acetal/Stainless Steel Thumbwheel, PTFE Ferrule (1/4 in.), M-2 Vespel™ Ferrule (1/4 in.), M-2 A Vespel Graphite Ferrule (6 mm)
Quantity 1 Ea.
Product Line Vespel
For Use With (Equipment) Thermal Desorption Tubes
Vespel is a trademark of E. I. du Pont de Nemours and Company