Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Vesphene™ III se Disinfectant Cleaner

Catalog No. 19415155 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-155 5 gal.
17-463-007 1 gal.
19-415-156 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-415-155 Supplier STERIS Supplier No. 647505
Add to Cart
Add to Cart
This item is not returnable. View return policy

This Phenolic Disinfectant is the latest generation of high pH disinfectant specifically formulated for the routine disinfection of hard, non-porous surfaces.

The Vesphene™ III se Disinfectant is specifically formulated for the routine disinfection of hard, non-porous surfaces. At a use dilution of rate of 0.8% v/v (1:128), the Vesphene™ III Disinfectants kill a broad spectrum of common bacteria and fungi. The alkaline Vesphene™ III Disinfectant formulation can be used alone or in rotation with the LpH™ III Disinfectants. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS™ Technical Services team for individualized application and validation assistance.

How Vesphene™ III se Disinfectant Works

The STERIS™ Vesphene™ III se Phenolic Disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.

Why Vesphene™ III se Disinfectant?

  • Efficacy
    • Phenolic active ingredients provide broad spectrum antimicrobial efficacy.
  • Productivity
    • Formulation includes surfactants for surface cleaning while disinfecting.
  • Flexibility
    • Ideal for use in a contamination control rotation.
  • Reliability
    • Complete lot traceability.
  • Assurance
    • Each lot documented by Certificate of Analysis.

Specifications

Product Type Phenolic Disinfectant
Form Concentrated Liquid
Sterility Non-sterile
Concentration 0.8% v/v (1:128)
Container Type Pail
For Use With (Application) Cleanroom, Controlled Environments
Volume (English) 5 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.