Learn More
Chemglass Life Sciences Vessel, Pressure, 200mL, #25 Thread, Round Bottom Flask, Vessel Only
SDP
Supplier:  Chemglass Life Sciences CG1880R12
Encompass_Preferred
Vessel, Pressure, 200mL, #25 Thread, Round Bottom Flask, Vessel Only. Heavy wall pressure vessel is the same as CG-1880, but shaped similar to a round bottom flask with a #25 internal thread for use with a PTFE bushing as a pressure seal. Complete apparatus is supplied with vessel, back seal solid PTFE bushing and viton o-ring.An optional Perfluoro o-ring has the chemical resistance and service temperature range similar to Kalrez 4079. See CG-362 and CG-366 for optional PTFE bushings.IMPORTANT NOTICE: ALL CHEMGLASS PRESSURE VESSELS ARE TESTED AT 150 PSI, OR APPROX TWO AND ONE-HALF TIMES THEIR RECOMMENDED WORKING PRESSURE. HOWEVER, CHEMGLASS DOES NOT GUARANTEE THE VESSEL FROM BREAKING UNDER PRESSURE, DUE TO THE NATURE OF MATERIAL AND CONDITIONS BEYOND OUR CONTROL. CHEMGLASS RECOMMENDS THE USE OF SAFETY SHIELDS AND/OR OTHER SAFETY EQUIPMENT.
Catalog No. 50-873-220
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.