Learn More
Red Kap™ ESD, Anti-Static Counter Lab Jackets
Catalog No. 19199005 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-199-005 3X-Large
19-199-002 Large
19-199-003 X-Large
19-199-001 Medium
19-199-000 Small
19-199-004 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-199-005 Supplier Red Kap™ Supplier No. KK26BL RG 3XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Electronic blue counter jacket features a five-gripper front as well as adjustable gripper sleeves with grounding snaps.
  • One-piece collar with notched lapel
  • Adjustable gripper sleeve with grounding snaps
  • Left chest pocket
  • Two lower pockets
Compliance
Meets ESD STM21-1997 standard
Specifications
Product Type General Purpose Lab Coat
Size 3X-Large
Gender Unisex
Material Carbon-modified Nylon
Color Electronic Blue
Closure Type Snaps
Disposable Reusable
Sterility Non-sterile
Resistance Static
Collar Type One-piece, Notched Lapel
Certifications/Compliance ANSI, ESD STM21-1997
Length (English) 33 in.
Length (Metric) 83.82 cm
Weight (English) 4 oz.
Weight (Metric) 113.39 g
Construction Polyester, Carbon Suffused Nylon