Learn More
Sterlitech Corporation VF17, GLASS FILTRATION APPARAT
SDP
Supplier:  Sterlitech Corporation 16720017
Encompass_Preferred
VF17, Glass Filtration Apparatus Includes 1000mL/90mm Glass Filter Holder, No. 8 Stopper, 3500mL Receiver Flask And GL45 Adaptor
Catalog No. NC2184913
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.