Learn More
Thermco Products VFC Temperature USB-C Data Logger
Catalog No. 04500340
Change view
Click to view available options
Display Type:
LCD
Catalog No. Display Type
04-500-340 LCD
1 options
Catalog No. 04-500-340 Supplier Thermco Products Supplier No. TRED3016CU
Add to Cart
Add to Cart
The CDC/VFC-compliant ISO/IEC 17025 NIST-traceable 30 day temperature data logger.
  • CDC and VFC compliant with a large, easy-to-read display designed for monitoring vaccine storage in refrigerators and freezers, meeting or exceeding CDC guidelines.
  • Stores over 17,500 temperature readings across a range of -40°C to +99°C (-40°F to +210°F).
  • Supports both smart probes and traditional probes.
  • Display shows current, minimum, and maximum temperatures, alarm status, and battery life for up to 59 days at a glance.
  • Pre-set alarm limits trigger visual alerts, changing the status indicator from ✓ to ✗, with optional audible alerts and day markers for temperature excursions.
  • Excursion details for the past 60 days can be reviewed directly on the display.
  • Data can be downloaded via a LogTag Interface cradle to LogTag Analyzer software for detailed analysis and reporting.
  • Key features include CDC compliance, large display with extended history, visual and audible alarms, and easy data download.
Specifications
Type Temperature Datalogger
Display Type LCD
Alarm Types Audible