Learn More
Grainger VGA to Svideo/RCA Adapter Cable - Black
Supplier:  Grainger 14C213
Encompass_Preferred
The Grainger Approved VGA to Svideo RCA Adapter allows you to connect a VGA source device such as a computer to an Svideo or RCA input on a TV or other display. It converts the VGA signal to Svideo and RCA outputs for compatibility with older TVs. This adapter supports resolutions up to 1600 x 1200 and has 15-pin HD male and female connectors. It measures 5 inches long and comes with a 1 year warranty from Grainger. This VGA to RCA/Svideo adapter provides an easy way to display a computer or other VGA source on a standard definition TV.
Catalog No. 50-275-2411
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.