Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567605 Supplier Invitrogen™ Supplier No. PA567605
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VGLL2 Polyclonal Antibody for Western Blot, IHC (P)

VGLL2 is the master regulator in the wing development in Drosophila, which acts through a complex by interacting with scalloped, a homolog of vertebrate TEF proteins. Northern and dot blot analyses of human adult tissues detected VGLL2 expression in skeletal muscle. In situ hybridization revealed VGLL2 expression in differentiating somites and branchial arches during mouse embryogenesis. VITO1 is the clone of VGLL2. It contains a scalloped interaction domain (SID). VITO1 is more highly expressed in differentiated myotubes from the mouse C2C12 myogenic cell line than in undifferentiated cells. During differentiation in mouse C2C12 cell line, VGLL2 mRNA levels increase and VGLL2 protein translocates from the cytoplasm to the nucleus. Interference with VITO1 expression by small interfering RNA attenuated differentiation in C2C12 muscle cells. Therefore, this protein is an important component of the myogenic program.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene VGLL2
Gene Accession No. Q8N8G2
Gene Alias C130057C21Rik; Protein VITO1; RP1-179P9.2; Transcription cofactor vestigial-like protein 2; Vestigial and Tondu related protein 1; vestigial like 2; vestigial like 2 homolog; vestigial like 2 homolog (Drosophila); vestigial like family member 2; vestigial-like 2; vestigial-like family member 2; Vgl2; vgl-2; Vgll2; Vito1; VITO-1
Gene Symbols VGLL2
Host Species Rabbit
Immunogen Full length Fusion protein of human VGLL2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245806
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.