Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VGLL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545925 Supplier Invitrogen™ Supplier No. PA545925
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HSGCAAPGPA SYRRPPSAAT TCDPVVEEHF RRSLGKNYKE PEPAPNSVSI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.

Transcription cofactor vestigial-like protein 4 (VGLL4) acts as a specific coactivator for the mammalian TEFs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGLL4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Vgll4
Gene Accession No. Q14135
Gene Alias BC048841; KIAA0121; Transcription cofactor vestigial-like protein 4; vestigial like 4; vestigial like 4 (Drosophila); vestigial like family member 4; Vestigial-like 4; vestigial-like family member 4; VGL-4; Vgll4
Gene Symbols Vgll4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human VGLL4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9686
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.