Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VH 032
GREENER_CHOICE

Catalog No. 777410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-741-0 10 mg
77-745-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-741-0 Supplier Tocris Bioscience™ Supplier No. 7774/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of VHL; blocks interaction of VHL and HIF-α

VH 032 is an inhibitor of E3 ubiquitin ligase VHL (Kd = 185 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. VH 032 treatment causes dose dependent upregulation of HIF-target genes including CA9, GLUT1 and PHD2 in different cell lines. Cell permeable.

Specifications

Chemical Name or Material (2 S,4 R)-1-(-2-Acetamido-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
CAS 1448188-62-2
Quantity 10 mg
Target Von Hippel Lindau
Molecular Formula C24H32N4O4S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.