Learn More
Medchemexpress LLC Vh032-cyclopropane-f | 2306193-99-5 | MFCD32201089 | 99.9% | 532.63 | C26H33FN4O5S | 25MG
SDP
Supplier:  Medchemexpress LLC HY12590525MG
Encompass_Preferred
VH032-cyclopropane-F is a VH032-based von Hippel-Lindau (VHL) ligand designed as an E3 ligase binder and building block for bifunctional degraders (PROTACs). It contains a functional handle that enables rapid conjugation to linkers, and is provided as a well-characterized, high-purity small molecule for chemical biology and targeted protein degradation research.
  • Functionalized VHL ligand suitable for PROTAC assembly.
  • Terminal hydroxyl group for easy linker conjugation.
  • High purity for reproducible biological studies.
  • Well-characterized molecular properties for design and synthesis.
  • Available in multiple small-scale quantities for research use.
Catalog No. 50-002-34945
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.