Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHH Rabbit anti-Llama, Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NB024714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024714 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB024714 Supplier Novus Biologicals Supplier No. NBP295273100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VHH Polyclonal specifically detects VHH in Llama samples. It is validated for ELISA/Western Blot.

Specifications

Antigen VHH
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene Alias anti-VhH, camelid VhH, single-domain antibody (sdAb)
Host Species Rabbit
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a VHH camelid domain protein.
Purification Method Sterile filtered
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Llama
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.