Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHL Mouse anti-Human, Clone:, BD Pharmingen™

Catalog No. BDB556347 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556347 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556347 Supplier BD Biosciences Supplier No. 556347
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VHL Monoclonal antibody specifically detects VHL in Human, Mouse samples. It is validated for Immunohistochemistry (Formalin-Antigen Retrieval Required) .
TRUSTED_SUSTAINABILITY

Specifications

Antigen VHL
Applications Immunohistochemistry
Classification Monoclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias von Hippel-Lindau protein; RCA1; HRCA1
Host Species Mouse
Immunogen Human VHL aa. 1-213 Recombinant Protein
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.