Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 1.8ml, Maximum Recovery, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Tan Silicone Rubber / White PTFE Septa bonded in the Blue Caps. 1/PK.
Supplier:  Microsolv Technology Corporation 9512S1CP
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
Catalog No. 50-227-0042