Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Vial Holder for HG-600 Geno/Grinder™ 2010 Homogenizer

Catalog No. 2197
Change view
Click to view available options
Length (Metric):
11.9 cm
Holds:
24 Standard 15 mL Conical Bottom Centrifuge Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Holds
2197 11.9 cm 24 Standard 15 mL Conical Bottom Centrifuge Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2197 Supplier Cole-Parmer™ Supplier No. 2197
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Vial Holder for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.

  • Safely grind samples in the HG-600 Geno/Grinder™ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
  • Order homogenizer and vials separately.

Specifications

Product Type Holder
For Use With (Equipment) HG-600 Geno/Grinder™ 2010 Homogenizer
Holds 24 Standard 15 mL Conical Bottom Centrifuge Tubes
Length (Metric) 11.9 cm
Material Foam
Packaging Quantity 1 Ea.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.