Learn More
Krackeler Scientific Vial Inserts
SDP
Catalog No. 056005204N
Change view
Click to view available options
Material:
Glass
Polypropylene
Product Type:
Inserto
Vial Insert with Polymer Feet
Volume (Metric) Insert:
150 μL
250 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Volume (Metric) Insert
056005204N Glass Vial Insert with Polymer Feet 150 μL
HP51811270N Glass Vial Insert with Polymer Feet 250 μL
51820549N Polypropylene Vial Insert with Polymer Feet 250 μL
056005203N Glass Inserto 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 056005204N Supplier Krackeler Scientific Supplier No. 9251832088EA
Add to Cart
Add to Cart
For microvolume sampling
Specifications
Material Glass
For Use With (Application) Microvolume sampling
Volume (English) Insert 0.005 oz.
Volume (Metric) Insert 150 μL
Base Style Conical
Product Type Vial Insert with Polymer Feet