Learn More
Krackeler Scientific Vial Inserts
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Application) | Microvolume sampling |
| Volume (English) Insert | 0.008 oz. |
| Volume (Metric) Insert | 250 μL |
| Base Style | Conical |
| Product Type | Vial Insert with Polymer Feet |