Learn More
Thomas Scientific 2D Cryovials, 5mL, Self Standing, External Threading, Sterile
SDP
Supplier:  Thomas Scientific 1186P26
Encompass
PolarSafe™ 2D tubes are the ideal consumable for rapid processing of biological samples. The embedded Datamatix code inserted in the bottom of each tube corresponds to the “Code 128” barcode featured on the body of the tube. They provide time-saving and accurate organization and tracking of valuable samples. Innovative “channels of evacuation” designed between the tube and the 2D chip allows fluid to easily drain The 2D tubes are offered in both 2 and 5mL sizes Available with internally or externally threaded caps
Catalog No. 50-146-1726
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.