Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Vibration Generator Set
SDPSCI_ED

Catalog No. S00656 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00656 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00656 Supplier United Scientific™ Supplier No. VWG002
Add to Cart
Add to Cart

United Scientific Vibration Generator Set enables a simple, yet complete demonstration of vibration and waves.

A comprehensive set of mechanical vibration and wave motion apparatus based on a sturdy, reliable vibration generator that can be used with any frequency generator that can deliver a signal of up to 1 A.

  • The piston delivers an amplitude of up to 7 mm, depending on the load and frequency.
  • The generator can be used on the bench or attached to stand material via a threaded socket on the bottom surface.
  • The accessories include a wire hoop and a set of vibrating metal strips for mechanical resonance studies as well as a spiral spring and an elastic cord for wave investigations.

Specifications

Grade 6 to 12
Format Kit
Languages English
Subject Physics - Wave Motion
WARNING: Cancer - www.P65Warnings.ca.gov

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.