Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Vibrations and Waves Set
SDPSCI_ED

Catalog No. S43681 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43681 5 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43681 Supplier United Scientific Supplies Supplier No. VWS001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Classroom demonstration of vibrations and waves

  • This comprehensive and economical set is based on a mechanical vibration generator and a sine wave signal generator and includes a support system and accessories.
  • The important basic types of mechanical vibrations and waves can be investigated: Transverse standing waves on a vibrating string, Longitudinal standing waves on a helical spring, Standing waves on a hoop (model of atomic quantization), Resonant vibrations of metal strips, Resonance patterns on round and square vibrating plates (Chladni plates).
  • Features: Experiments are performed on a stable and easy-to-use extruded bench, Robust and reliable mechanical generator, Amplified sine wave generator with large, 3-digit frequency display, Two frequency ranges: 1—400 Hz x 1 Hz, 0.01—4.00 kHz x 0.01 kHz, easily visible results.
  • The wide range of accessories allows both of the fundamental topics of transverse and longitudinal waves to be demonstrated and enables further investigation of resonance phenomena in various systems from the famous and basic waves-on-a-string to advanced and complex nodal patterns on vibrating plates.

Specifications

Grade 5 to 15
Format Kit
Product Type Vibrations Set
Languages English
Subject Physics - Waves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.