Learn More
ATCC Vibrio campbellii; Strain KM413
Supplier:  ATCC BAA2363
Encompass
Vibrio campbellii strain KM413 is a whole-genome sequenced strain that was derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This culture is one of several strains included in a “quorum sensing kit” (ATCC MP-6) developed by the depositor. This mutant strain is deficient at autoinducer production but proficient at bioluminescence when provided with exogenous autoinducers.
Catalog No. 50-238-3736
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.