Learn More
ATCC Vibrio campbellii; BB152
Supplier:  ATCC BAA1119
Encompass
Vibrio campbellii strain BB152 is a whole-genome sequenced bacterium that was derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This culture is one of several strains included in a “quorum sensing kit” (ATCC MP-6) developed by the depositor. This strain has the genotype luxM::tn5Kan. It is a mutant sensor strain that responds only to the AI-2 autoinducer.
Catalog No. 50-238-0311
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.