Learn More
ATCC Vibrio cholerae
Supplier:  ATCC 14033
Encompass
suggested neotype. Nontoxigenic. Control strain for detection of Vibrio cholerae and V. vulnificus.
Catalog No. NC1647467
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.