Learn More
Creative Biomart Influenza A virus H3N2 (Brisbane) Antigen
Supplier:  Creative Biomart VAC0271
Encompass_Preferred
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza A virus, strain A/Brisbane/10/07. The Influenza Virus was purified by Ultracentrifugation with 10-40 % sucrose gradient.
Catalog No. 50-208-5111
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.