Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Victoria Blue B, pure

Catalog No. AC190191000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190191000 100 g Glass bottle
AC190190250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190191000 Supplier Thermo Scientific Chemicals Supplier No. 190191000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2580-56-5 | C33H32ClN3 | 506.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2580-56-5
Molecular Formula C33H32ClN3
Molecular Weight (g/mol) 506.09
MDL Number MFCD00011878
InChI Key LLWJPGAKXJBKKA-UHFFFAOYSA-N
Synonym Basic Blue 26, C.I. 44045
PubChem CID 17407
ChEBI CHEBI:87651
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]naphthalen-1-ylidene]-phenylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[NH+]C3=CC=CC=C3)C4=CC=CC=C24)C5=CC=C(C=C5)N(C)C.[Cl-]

Specifications

Melting Point 206.0°C
Infrared Spectrum Authentic
Lambda Max 599 to 605nm (in ethanol)
Loss on Drying 5% max. (60°C)
Packaging Glass bottle
UV Spectrometry Complies
Color Brown
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
Quantity 100 g
Physical Form Glistening Crystalline Powder
Formula Weight 506.09
Grade Pure
Chemical Name or Material Victoria blue B
Show More Show Less
EINECSNumber 219-943-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.