Learn More
Apexbio Technology LLC Vidarabine monohydrate 24356-66-9 50mg
SDP
Supplier:  Apexbio Technology LLC C637750
Encompass_Preferred
Vidarabine monohydrate (CAS 24356-66-9) is a small-molecule inhibitor targeting viral DNA polymerase It is designed to selectively inhibit viral DNA synthesis thereby disrupting viral DNA replication Vidarabine monohydrate exerts its biological activity primarily through incorporation into viral DNA strands causing premature DNA chain termination In in vitro studies Vidarabine monohydrate demonstrates antiviral activity with an IC50 range of approximately 3 to 10 M against herpes simplex viruses Based on these pharmacological properties Vidarabine monohydrate holds research potential in investigations of viral replication mechanisms antiviral drug resistance and DNA polymerase inhibition assays
Catalog No. 50-000-92793