Learn More
Vidaro™ C3™ B-Fore Cleanroom Coveralls
Catalog No. 19999838 Shop All Vidaro Corporation Products
Change view
Click to view available options
Color:
Light Blue
Navy
White
Size:
Medium
2X-Large
3X-Large
4X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-999-838 Navy 4X-Large
19-999-820 White 2X-Large
19-999-821 White 3X-Large
19-999-822 White 4X-Large
19-999-828 Light Blue 2X-Large
19-999-830 Light Blue 4X-Large
19-999-829 Light Blue 3X-Large
19-999-825 Light Blue Medium
19-999-837 Navy 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-999-838 Supplier Vidaro™ Supplier No. C43587/4XL
This item is not returnable. View return policy
Cleanroom coveralls minimize particulate contamination and dissipate electrostatic charges.
Assure contamination control and increase worker comfort with reusable apparel made from this state-of-the-art, high-performance ESD cleanroom fabric. Designed for the microelectronics industry and any applications where products must be protected from static dissipative charges, airborne particles, and lint. With a super-dense weave of 100% microdenier polyester fibers (over 19,000 filaments per square inch) Burlington™ C-3 meets the highest standard of particulate control. A microengineered carbon fiber grid dissipates electrostatic charges and minimizes manufacturing defects. Made in USA.
  • Made of B-Fore fabric, a super-dense weave of 100% microdenier polyester fibers that minimize particulate contamination and a microengineered carbon fiber grid that dissipates electrostatic charges
  • Feature a convenient color-coded hanger loop for quick size identification
  • With military collar and long raglan sleeves with ESD knit cuffs
  • Full-length, limited-linting front zipper with two-ply fly stiffener closure, and snap adjustments at the collar and ankles
  • Snaps on back of each leg attach to boots and prevent pullout
Order Info
Available in white,light blue and navy. Made in USA.
Specifications
Product Type Cleanroom Coveralls
Size 4X-Large
Material Polyester
Color Navy
Disposable Reusable
Closure Type Zipper