Learn More
Vidaro™ B-Fore™ ESD Hoods
Catalog No. 19999883 Shop All Vidaro Corporation Products
Change view
Click to view available options
Color:
Light Blue
Navy
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-999-883 White X-Large
19-999-860 Light Blue 2X-Large
19-999-873 Navy X-Small
19-999-874 Navy Small
19-999-875 Navy Medium
19-999-876 Navy Large
19-999-877 Navy X-Large
19-999-878 Navy 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-999-883 Supplier Vidaro™ Supplier No. H3158/XL
This item is not returnable. View return policy
Requires
Two female snaps on each cheek allow the attachment of a facial (not included).
High-performance fabric ideal for critical environments.
  • Super-dense weave of 100% microdenier polyester fibers minimizes particulate contamination
  • Microengineered carbon fiber grid dissipates electrostatic charges
  • Open-faced hood fits snugly under the chin, leaving the entire face exposed
  • Full-bib apron is long, to fit securely into a coverall or frock without pulling out.
  • Front closure has two stainless-steel snaps, and there are adjustment snaps at the back of the neck
Specifications
Product Type Open Face
Size X-Large
Material Polyester, Carbon
Color White
Closure Type Snap Front
Disposable Reusable
Model H-3158
Quantity 1 Ea.
Includes Two snaps on each cheek, Stainless Steel front-snap closure, snap at back of neck for adjustment