Learn More
Vidaro™ Unisex Reusable Cotton Lab Coats
Catalog No. 19063800 Shop All Vidaro Corporation Products
Change view
Click to view available options
Style:
Long
Short
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
19-063800 Short
19-063826 Long
19-063834 Long
19-063859 -
19-021295 Long
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-063800 Supplier Vidaro™ Supplier No. L213H/40 (S/12)
This item is not returnable. View return policy
White lab coats with reinforced side slits and plastic button closure are available in short or long styles.
  • Reinforced side-slits for access to inner garment
  • 7 oz./square yard 100% cotton twill
  • Two-ply laydown collar and lapel
  • Plastic-button front closure
  • Comes in a variety of sizes
  • Bar-tacked at all stress points
  • Long sleeve with banded cuff for durability
  • Banded back with pleats
  • Left breast pocket with pencil slot
  • Two lower front pockets
Specifications
Product Type Controlled Environments Lab Coat
Gender Unisex
Disposable Reusable
Style Short
No. of Pockets 3
Model L-213-K
Size (English) Hip 30 in.
Size (Metric) Hip 76.2 cm