Description
- Open face polyester cleanroom hoods
- Sizes range from x-small to 2X-large
- Polyester material
- Two snap closure on each cheek
- Stainless steel front-snap closure
- Adjustable back snap at neck
Specifications
Specifications
| Product Type | Open Face |
| Size | X-Small |
| Material | Polyester |
| Color | White |
| Closure Type | Snap Front |
| Disposable | Single-use |
| Model | H-3111 |
| Quantity | 1 Ea. |
| Includes | Two snaps on each cheek, Stainless Steel front-snap closure, adjustable back snap at neck |