Learn More
Syngene™ Viewing Platform
GREENER_CHOICESDP
Catalog No. 01257235 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
GBOX Mini System
Catalog No. For Use With (Equipment)
01-257-235 GBOX Mini System
1 options
Catalog No. 01-257-235 Supplier Syngene™ Supplier No. GEVP
Add to Cart
Add to Cart
Viewing platform for use with GBOX Mini system.
  • Designed to be used instead of a transilluminator
Specifications
Product Line Syngene
For Use With (Application) Providing white light under a sample
Product Type Viewing Platform
For Use With (Equipment) GBOX Mini System
Voltage 100/115/220/240 V
WARNING: Cancer - www.P65Warnings.ca.gov