Learn More
Greiner Bio-One VIEWseal Adhesive Microplate Sealer
GREENER_CHOICE
Catalog No. 07000408 Shop All Greiner Bio-One Products
Change view
Click to view available options
Adhesive:
Silicone
Catalog No. Adhesive
07-000-408 Silicone
1 options
Catalog No. 07-000-408 Supplier Greiner Bio-One Supplier No. 676070
Add to Cart
Add to Cart
For protein crystallography. Greiner Bio-One VIEWseal Adhesive Microplate Sealer is special highly transparent sealer with low autofluorescence for precise optical measurements.
  • Coated with silicone adhesive
  • Non-Pyrogenic
  • Highly transparent film for optical measurements
  • Sticks only when film surface is pressed on
  • Film does not adhere to gloves
  • Temperature-resistant from -80°C to
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Sterility Non-sterile
No. per Case 100
Product Type Sealer
Adhesive Silicone
Plate Compatibility Microplate
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.