Learn More
United Scientific Supplies Vigreux Distilling Columns, 24/40 Joint
SDPSCI_ED
Catalog No. S43792 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
200 mm
300 mm
400 mm
500 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S43792 200 mm
S43793 300 mm
S43794 400 mm
S43795 500 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S43792 Supplier United Scientific Supplies Supplier No. CDV200
Add to Cart
Add to Cart
Allows for a dramatic increase in surface area per unit length
  • Vigreux columns have a modified sample tube to include an abundance of downward-pointing indentations
  • This design dramatically increases the surface area per unit length of the condenser
  • Includes a 24/40 connector
Specifications
For Use With (Equipment) Air Condenser
Material Borosilicate Glass
Connection Type 24/40
Type Vigreux Distilling Column
For Use With (Application) Distillation
Length (Metric) 200 mm