Learn More
United Scientific Supplies Vigreux Distilling Columns, 24/40 Joint
Shop All United Scientific Supplies, Inc Products
Description
- Vigreux columns have a modified sample tube to include an abundance of downward-pointing indentations
- This design dramatically increases the surface area per unit length of the condenser
- Includes a 24/40 connector
Specifications
Specifications
| For Use With (Equipment) | Air Condenser |
| Material | Borosilicate Glass |
| Connection Type | 24/40 |
| Type | Vigreux Distilling Column |
| For Use With (Application) | Distillation |
| Length (Metric) | 300 mm |