Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ 0.6 in. Drain Cleaning Brush

Catalog No. 17227379 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-379 Black
17-227-378 Red
17-227-377 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-227-379 Supplier Vikan™ Supplier No. 53609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vikan™ 0.6 in. Drain Cleaning Brush features stiff bristles mounted on a flexible stainless steel rod.

  • Used to clean small spaces between machine parts and drainage holes in sinks
  • The short handle allows the operator to exert pressure to get drains clean
  • The stiff bristles back the effort up
  • Color-coded

Specifications

Product Type Cleaning Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Black
Diameter (English) Bristle 0.6 in.
Length (English) 12.2 in.
Quantity 1 Ea.
Size Small
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.