Description
- Designed to clean hard-to-reach areas
- Soft bristles are attached to a helically wound stainless steel shaft
- Strong, FDA-compliant, and chemically resistant
- The brush can also be used with water and a cleaning agent for a washing effect
- Color-coded
Specifications
Specifications
| Product Type | Tube Brush |
| Color-coded | Yes |
| Handle Material | Stainless Steel |
| Sterility | Non-sterile |
| Certifications/Compliance | FDA Compliant |
| Color | White |
| Length (English) | 36 in. |
| Quantity | 1 Ea. |
| Size | Medium |