Description
- Suitable for cleaning with the use of aggressive chemicals and where the handle is exposed to a heavy-handed treatment
- The hygiene sleeve makes the handle more ergonomically friendly
- Color-coded
Specifications
Specifications
| Color Code | Green |
| Product Type | Handle |
| Sterility | Non-sterile |
| Quantity | 1 Ea. |
| Description | Thread type Euro Hygienic |
| Color-coded | Yes |
| Material | Stainless Steel |