Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ 5 in. Medium Pipe Brush

Catalog No. 17227752 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-752 Red
17-227-754 Yellow
17-227-750 Green
17-227-751 Blue
17-227-753 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-752 Supplier Vikan™ Supplier No. 70354
Add to Cart
Add to Cart

Vikan™ 5 in. Medium Pipe Brush can be used to clean long pipes, high walls, and tanks.

  • Medium bristles on this brush make it useful to clean areas like small tanks and containers
  • Can be used to clean mixers for blending ingredients
  • Fits well into meat mincer barrels
  • Requires a European-thread handle
  • Can be attached to any length handle to fit the user's needs
  • Medium bristles can be used dry to sweep off tough dirt
  • Brush can be combined with water and a cleaning agent for scrubbing
  • Color-coded

Specifications

Product Type Pipe Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Red
Description Thread type Euro Hygienic
Length (English) 4.4 in.
Quantity 1 Ea.
Size Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.