Learn More
Description
- For use with pad holder and deck scrubs
- Suitable for reaching high walls and tanks
- Cna be used with clean-in-place brushes for cleaning on top of overhead pipes
- The handle extends to a maximum length of 115 in.
- Uniquely suited for color-coding and HACCP programs
Specifications
Specifications
| Product Type | Handle |
| Color Code | Green |
| Sterility | Non-sterile |
| Quantity | 1 Ea. |
| Description | Thread type Euro Hygienic |
| Certifications/Compliance | FDA Compliant |
| Color-coded | Yes |
| Material | Aluminum, Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.