Description
- Quick connect coupling with shut-off valve
- Combines a water hose and any of our waterfed products to ensure a watertight fit
- Bottom piece slides down and up to both connect and disconnect while shutting off the water supply
- Color-coded
Specifications
Specifications
| Product Type | Coupling |
| Color | Silver |
| Material | Chrome-plated Brass |
| Quantity | 1 |
| Dimensions (L x W x H) | 1.95 x 1.02 x 1.95 in. |