Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Basic Wall Bracket

Catalog No. 17227008 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Red
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-008 Black
17-227-003 Red
17-227-006 Orange
17-227-005 Yellow
17-227-001 Green
17-227-002 Blue
17-227-004 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-227-008 Supplier Vikan™ Supplier No. 06159
Add to Cart
Add to Cart

Vikan™ Color-Coded Basic Wall Bracket is a one-piece wall bracket with mounting kit that can hold up to three products.

  • Supports any product and is virtually unbreakable
  • Includes screws, wall anchors, and mounting instructions
  • Provides a color-coded storage option for tools
  • Proper storage of cleaning equipment
  • Supports hygienic environments
  • Helps to extend the usable life of the equipment

Specifications

Product Type Wall Bracket
Color Black
Material Polypropylene
For Use With (Application) Color-coded storage
Certifications/Compliance FDA Compliant
Quantity 1
Dimensions (L x W x H) 9.4 x 6.5 x 2.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.