Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Stainless Steel Floor Scrapers

Catalog No. 17227052 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-052 Green
17-227-054 Red
17-227-056 Yellow
17-227-053 Blue
19-830-239 Black
17-227-055 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-227-052 Supplier Vikan™ Supplier No. 29102

Color-coded stainless steel floor scrapers can be combined with any Euro thread handle to get to hard-to-reach areas.

  • Ideal for removing stubborn debris and other substances
  • Long-lasting blade
  • Connects with any Remco™/Vikan™ handles to provide an easy solution for stuck-on messes
  • Scapper is hygienically designed for easy cleaning

Specifications

Product Type Floor Scraper
Color-coded Yes
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Green
Dimensions (L x W) 10.25 x 1.25 in.
For Use With (Application) Cleaning hard-to-reach areas
Material Polypropylene, Stainless Steel
Quantity 1 Ea.
Size Large
Weight (English) 0.56 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.