Learn More
Description
- Proper storage of cleaning equipment enhances food safety and extends the shelf life of your equipment
- These wall brackets can adjust to any color-coded program, and you can extend the length of the bracket system by mounting multiple brackets together adjacently
- The wall bracket comes with four rubber clips and two side hooks
- The rubber hooks and clips are interchangeable to adapt to custom storage needs
- The rubber clip design of this bracket formly holds equipment in place
Specifications
Specifications
| Product Type | Wall Bracket |
| Color | Green |
| Material | Polypropylene |
| For Use With (Application) | Storing equipment |
| Certifications/Compliance | FDA Compliant |
| Quantity | 1 |
| Dimensions (L x W x H) | 15.3 x 2.25 x 2.25 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.