Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Wall Bracket for 4 to 6 Tools

Catalog No. 17227034 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-034 Orange
17-227-031 Red
17-227-033 Yellow
17-227-029 Green
17-227-030 Blue
17-227-035 Purple
17-227-028 Pink
17-227-032 White
17-227-036 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-227-034 Supplier Vikan™ Supplier No. 10187

Vikan™ Color-Coded Wall Bracket for 4 to 6 Tools can be extended by mounting multiple pieces together adjacently.

  • Proper storage of cleaning equipment enhances food safety and extends the shelf life of your equipment
  • These wall brackets can adjust to any color-coded program, and you can extend the length of the bracket system by mounting multiple brackets together adjacently
  • The wall bracket comes with four rubber clips and two side hooks
  • The rubber hooks and clips are interchangeable to adapt to custom storage needs
  • The rubber clip design of this bracket formly holds equipment in place

Specifications

Product Type Wall Bracket
Color Orange
Material Polypropylene
For Use With (Application) Storing equipment
Certifications/Compliance FDA Compliant
Quantity 1
Dimensions (L x W x H) 15.3 x 2.25 x 2.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.