Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Wall Bracket Replacement Clips

Catalog No. 17227045 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-045 Black
17-227-040 Red
17-227-043 Orange
17-227-042 Yellow
17-227-038 Green
17-227-039 Blue
17-227-044 Purple
17-227-037 Pink
17-227-041 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-227-045 Supplier Vikan™ Supplier No. 10199

Vikan™ Color-Coded Wall Bracket Replacement Clips combine with a wall bracket system to allow for total customization of your storage systems.

  • Contains 4 rubber clips and 8 pins
  • For use with wall brackets 1017 and 1018
  • Allows total customization of the wall bracket system to fit any color-coded program
  • Extra clips can be added to hold more tools with any Vikan™/Remco™ handles

Specifications

Product Type Wall Bracket
Color Black
Material Rubber, Polypropylene
For Use With (Application) Wall bracket customization
Certifications/Compliance FDA Compliant
Quantity 1
Dimensions (L x W x H) 4.7 x 3.9 x 1.6 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.