Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Color-Coded Wall Bracket Replacement Hooks

Catalog No. 17227011 Shop All Remco Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-011 Green
17-227-013 Red
17-227-016 Orange
17-227-015 Yellow
17-227-012 Blue
17-227-017 Purple
17-227-010 Pink
17-227-014 White
17-227-018 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-227-011 Supplier Vikan™ Supplier No. 10162

Vikan™ Color-Coded Wall Bracket Replacement Hooks combine with a wall bracket system to allow for total customization of your storage systems.

  • Contains 4 hooks and 8 pins
  • For use with wall brackets 1017 and 1018
  • Allows total customization of the wall bracket system to fit any color-coded program
  • Extra hooks can be added to hold more hand tools such as brushes, scrapers, and squeegees
  • Hold more smaller hand tools with these easy-to-install hooks

Specifications

Product Type Hooks
Color Green
Material Polypropylene
For Use With (Application) Customizing wall bracket storage systems
Certifications/Compliance FDA Compliant
Quantity 1
Dimensions (L x W x H) 3 x 0.8 x 2.4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.