Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Extra Stiff X-Large Hand Brush

Catalog No. 17227236 Shop All Remco Products
Change view
Click to view available options
Color:
Green
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-236 Yellow
17-227-232 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-227-236 Supplier Vikan™ Supplier No. 38926

missing translation for 'validMainframeMsg'

Vikan™ Extra Stiff X-Large Hand Brush is designed to clean large surfaces and tough messes efficiently.

  • With flared bristles
  • For cleaning tables, chopping boards, buckets, large bowls, and other equipment
  • Ideal for reaching into corners
  • Large size makes it easy to clean
  • Color-coded

Specifications

Product Type Hand Brush
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Yellow
Length (English) 9.8 in.
Quantity 1 Ea.
Size X-Large
Width (English) Bristle 4.75 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.