Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ Flexible Paddle Scraper

Catalog No. 17227746 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-746 Blue
17-227-747 Red
17-227-749 Yellow
17-227-745 Green
17-227-748 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-227-746 Supplier Vikan™ Supplier No. 70133
Add to Cart
Add to Cart

Flexible paddle scraper is used to remove remaining food stuff from containers prior to the cleaning procedure.

  • Intended for scraping substances from the sides of containers
  • Ideal tool for prepping prior to the cleaning procedure
  • Euro-thread connection allows for pairing with any length or type of Vikan™ handle
  • Can be used on everything from large vats to simple mixing bowls
  • Color-coded

Specifications

Product Type Scraper
Color-coded Yes
Color Code Blue
Certifications/Compliance FDA Compliant
Color Blue
Material Polyethylene
Size Medium
Description Thread type Euro Hygienic
Dimensions (L x W) 8.75 x 4.3 in.
Quantity 1 Ea.
Includes Thread type Euro Hygienic
Weight (English) 0.26 lb.
No. per Pack 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.